The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257386.1 | NP_001244315.1 | 5 - 6 | 882 | 92 |
NM_004827.2 | NP_004818.2 | 5 - 6 | 1027 | 92 | |
XM_005263354.3 | XP_005263411.1 | 5 - 6 | 870 | 92 | |
XM_005263355.3 | XP_005263412.1 | 5 - 6 | 977 | 92 | |
XM_005263356.3 | XP_005263413.1 | 5 - 6 | 753 | 92 | |
XM_011532420.2 | XP_011530722.1 | 7 - 8 | 1141 | 92 | |
XM_017008852.1 | XP_016864341.1 | 5 - 6 | 870 | 92 | |
XM_017008853.1 | XP_016864342.1 | 5 - 6 | 667 | 92 | |
GenBank mRNA | AB051855.1 | - | 5 - 6 | 578 | 92 |
AB056867.1 | - | 4 - 5 | 534 | 92 | |
AF098951.2 | - | 5 - 6 | 772 | 92 | |
AF103796.1 | - | 5 - 6 | 738 | 92 | |
AF463519.1 | - | 4 - 5 | 542 | 92 | |
AK002040.1 | - | 2 - 3 | 222 | 92 | |
AK290000.1 | - | 5 - 6 | 598 | 92 | |
AY017168.1 | - | 5 - 6 | 727 | 92 | |
AY288307.1 | - | 2 - 3 | 192 | 92 | |
AY289766.1 | - | 5 - 6 | 734 | 92 | |
AY333755.1 | - | 4 - 5 | 543 | 92 | |
AY333756.1 | - | 4 - 5 | 543 | 92 | |
BC021281.2 | - | 5 - 6 | 1027 | 92 | |
BC092408.1 | - | 5 - 6 | 882 | 92 |