Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ING1
Entrez Gene ID
3621
Gene Name
inhibitor of growth family member 1
Gene Aliases
p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a
Chromosome Location
Chr.13: 110712623 - 110721074 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005537.5 NP_005528.4 1 - 2 998 92
GenBank mRNA AB024402.1 - 1 - 2 1027 92
AB031269.1 - 1 - 2 442 92
AF001954.1 - 1 - 2 197 92
AF181849.1 - 1 - 2 998 92
BC018348.1 - 1 - 2 433 92

View Full Product Details