Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSSC4
Entrez Gene ID
10078
Gene Name
tumor suppressing subtransferable candidate 4
Gene Aliases
-
Chromosome Location
Chr.11: 2400409 - 2403878 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297658.1 NP_001284587.1 3 - 4 778 99
NM_001297659.1 NP_001284588.1 2 - 3 626 99
NM_001297660.1 NP_001284589.1 3 - 4 610 99
NM_005706.3 NP_005697.2 2 - 3 469 99
XM_006718118.2 XP_006718181.1 2 - 3 1508 99
XM_011519830.2 XP_011518132.1 3 - 4 720 99
GenBank mRNA AF125568.1 - 1 - 2 159 99
BC006091.1 - 2 - 3 182 99
BI084274.1 - 2 - 3 627 98
BI768923.1 - 3 - 4 502 99
BX384913.2 - 3 - 4 339 99

View Full Product Details