The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135024.1 | NP_001128496.1 | 2 - 3 | 438 | 89 |
NM_001166158.1 | NP_001159630.1 | 2 - 3 | 767 | 89 | |
NM_003477.2 | NP_003468.2 | 2 - 3 | 767 | 89 | |
XM_011520390.1 | XP_011518692.1 | 2 - 3 | 191 | 89 | |
GenBank mRNA | AF001437.1 | - | 2 - 3 | 249 | 89 |
AK301384.1 | - | 2 - 3 | 248 | 89 | |
AK312463.1 | - | 2 - 3 | 263 | 89 | |
BC010389.1 | - | 2 - 3 | 271 | 89 | |
BI915296.1 | - | 2 - 3 | 115 | 89 | |
BX648910.1 | - | 2 - 3 | 325 | 89 | |
DA573433.1 | - | 2 - 3 | 440 | 89 | |
U82328.1 | - | 2 - 3 | 279 | 89 | |
Y13145.1 | - | 2 - 3 | 241 | 89 |