The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003480.3 | NP_003471.1 | 6 - 7 | 482 | 82 |
NR_123733.1 | 6 - 7 | 482 | 82 | ||
GenBank mRNA | AK290585.1 | - | 6 - 7 | 273 | 82 |
AK315807.1 | - | 6 - 7 | 269 | 82 | |
AU280458.1 | - | 6 - 7 | 420 | 82 | |
BC005901.1 | - | 6 - 7 | 272 | 82 | |
DN996275.1 | - | 6 - 7 | 303 | 82 | |
U37283.1 | - | 6 - 7 | 404 | 82 |