The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003562.4 | NP_003553.2 | 1 - 2 | 351 | 92 |
GenBank mRNA | AF070548.1 | - | 1 - 2 | 205 | 92 |
AK290376.1 | - | 1 - 2 | 102 | 92 | |
AK313010.1 | - | 1 - 2 | 207 | 92 | |
BC006508.2 | - | 1 - 2 | 207 | 92 | |
BC006519.2 | - | 1 - 2 | 207 | 92 | |
BC016294.2 | - | 1 - 2 | 126 | 92 | |
BC017170.2 | - | 1 - 2 | 207 | 92 | |
CB155476.1 | - | 1 - 2 | 351 | 92 | |
CR456834.1 | - | 1 - 2 | 94 | 92 |