Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45217450-45217956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L2
Entrez Gene ID
90332
Gene Name
exocyst complex component 3 like 2
Gene Aliases
XTP7
UniGene
Hs.337557
Gene Chromosome Location
Chr.19: 45212621-45234211 on Build GRCh38
Transcripts
NM_138568.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5365698 c.760C>T p.R254C Chr.19: 45217587-45217587 on Build GRCh38
6026223 c.792G>A p.A264A Chr.19: 45217555-45217555 on Build GRCh38
4778405 c.780G>C p.R260R Chr.19: 45217567-45217567 on Build GRCh38
6026224 c.747G>T p.A249A Chr.19: 45217600-45217600 on Build GRCh38
4803941 c.775C>T p.L259F Chr.19: 45217572-45217572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905267483 Chr.19: 45217850-45217850 on Build GRCh38
rs927665393 Chr.19: 45217662-45217662 on Build GRCh38
rs768964166 Chr.19: 45217594-45217594 on Build GRCh38
rs910949198 Chr.19: 45217764-45217764 on Build GRCh38
rs751711942 Chr.19: 45217867-45217867 on Build GRCh38
rs373195270 Chr.19: 45217691-45217691 on Build GRCh38
rs763798851 Chr.19: 45217555-45217555 on Build GRCh38
rs946591263 Chr.19: 45217721-45217721 on Build GRCh38
rs1050220563 Chr.19: 45217725-45217725 on Build GRCh38
rs768898026 Chr.19: 45217479-45217479 on Build GRCh38
rs915129937 Chr.19: 45217716-45217716 on Build GRCh38
rs993890503 Chr.19: 45217775-45217775 on Build GRCh38
rs905377252 Chr.19: 45217706-45217706 on Build GRCh38
rs897951988 Chr.19: 45217910-45217910 on Build GRCh38
rs776755078 Chr.19: 45217487-45217487 on Build GRCh38
rs912558073 Chr.19: 45217739-45217739 on Build GRCh38
rs564783215 Chr.19: 45217532-45217532 on Build GRCh38
rs747364981 Chr.19: 45217584-45217584 on Build GRCh38
rs761874137 Chr.19: 45217495-45217495 on Build GRCh38
rs1050500859 Chr.19: 45217490-45217490 on Build GRCh38
rs529408246 Chr.19: 45217735-45217735 on Build GRCh38
rs889195876 Chr.19: 45217501-45217501 on Build GRCh38
rs1045195768 Chr.19: 45217843-45217843 on Build GRCh38
rs769916286 Chr.19: 45217498-45217498 on Build GRCh38
rs972852540 Chr.19: 45217743-45217743 on Build GRCh38
rs759472317 Chr.19: 45217536-45217536 on Build GRCh38
rs971528858 Chr.19: 45217796-45217796 on Build GRCh38
rs1023148348 Chr.19: 45217788-45217788 on Build GRCh38
rs983848421 Chr.19: 45217763-45217763 on Build GRCh38
rs952273405 Chr.19: 45217616-45217616 on Build GRCh38
rs1027937837 Chr.19: 45217784-45217784 on Build GRCh38
rs773309224 Chr.19: 45217686-45217686 on Build GRCh38
rs893818001 Chr.19: 45217791-45217791 on Build GRCh38
rs1013139067 Chr.19: 45217787-45217787 on Build GRCh38
rs760428317 Chr.19: 45217551-45217551 on Build GRCh38
rs897848016 Chr.19: 45217767-45217767 on Build GRCh38
rs1052746063 Chr.19: 45217790-45217790 on Build GRCh38
rs2109474 Chr.19: 45217506-45217506 on Build GRCh38
rs371811745 Chr.19: 45217568-45217568 on Build GRCh38
rs946643557 Chr.19: 45217901-45217901 on Build GRCh38
rs780478074 Chr.19: 45217582-45217582 on Build GRCh38
rs549454046 Chr.19: 45217756-45217756 on Build GRCh38
rs537383499 Chr.19: 45217518-45217518 on Build GRCh38
rs767177007 Chr.19: 45217542-45217542 on Build GRCh38
rs957165734 Chr.19: 45217649-45217649 on Build GRCh38
rs758971752 Chr.19: 45217577-45217577 on Build GRCh38
rs567037831 Chr.19: 45217729-45217729 on Build GRCh38
rs752559250 Chr.19: 45217543-45217543 on Build GRCh38
rs969129812 Chr.19: 45217634-45217634 on Build GRCh38
rs988205353 Chr.19: 45217870-45217870 on Build GRCh38
rs753604610 Chr.19: 45217556-45217556 on Build GRCh38
rs1018908216 Chr.19: 45217511-45217511 on Build GRCh38
rs778574363 Chr.19: 45217566-45217566 on Build GRCh38
rs760505176 Chr.19: 45217731-45217731 on Build GRCh38
rs981876935 Chr.19: 45217840-45217840 on Build GRCh38
rs916375346 Chr.19: 45217633-45217633 on Build GRCh38
rs981826033 Chr.19: 45217638-45217638 on Build GRCh38
rs914872632 Chr.19: 45217873-45217873 on Build GRCh38
rs934969192 Chr.19: 45217677-45217677 on Build GRCh38
rs547242531 Chr.19: 45217668-45217668 on Build GRCh38
rs769797429 Chr.19: 45217648-45217648 on Build GRCh38
rs756771513 Chr.19: 45217560-45217560 on Build GRCh38
rs1028302749 Chr.19: 45217578-45217578 on Build GRCh38
rs988127786 Chr.19: 45217734-45217734 on Build GRCh38
rs944804830 Chr.19: 45217730-45217730 on Build GRCh38
rs942564830 Chr.19: 45217773-45217773 on Build GRCh38
rs1052386684 Chr.19: 45217711-45217711 on Build GRCh38
rs867150036 Chr.19: 45217914-45217914 on Build GRCh38
rs962325003 Chr.19: 45217533-45217533 on Build GRCh38
rs991650719 Chr.19: 45217619-45217619 on Build GRCh38
rs569214026 Chr.19: 45217785-45217785 on Build GRCh38
rs79186375 Chr.19: 45217866-45217866 on Build GRCh38
rs952447949 Chr.19: 45217759-45217759 on Build GRCh38
rs79187950 Chr.19: 45217539-45217539 on Build GRCh38
rs772022601 Chr.19: 45217710-45217710 on Build GRCh38
rs993773362 Chr.19: 45217917-45217917 on Build GRCh38
rs537864887 Chr.19: 45217823-45217823 on Build GRCh38
rs748379143 Chr.19: 45217602-45217602 on Build GRCh38

View Full Product Details