Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45231656-45232162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L2
Entrez Gene ID
90332
Gene Name
exocyst complex component 3 like 2
Gene Aliases
XTP7
UniGene
Hs.337557
Gene Chromosome Location
Chr.19: 45212621-45234211 on Build GRCh38
Transcripts
NM_138568.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3389133 c.7G>A p.A3T Chr.19: 45231846-45231846 on Build GRCh38
7348063 c.76G>A p.V26I Chr.19: 45231777-45231777 on Build GRCh38
6880881 c.44C>T p.P15L Chr.19: 45231809-45231809 on Build GRCh38
372949 c.43C>A p.P15T Chr.19: 45231810-45231810 on Build GRCh38
3764751 c.86T>C p.V29A Chr.19: 45231767-45231767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756625243 Chr.19: 45231844-45231844 on Build GRCh38
rs757415310 Chr.19: 45231778-45231778 on Build GRCh38
rs996097941 Chr.19: 45231965-45231965 on Build GRCh38
rs954559390 Chr.19: 45231687-45231687 on Build GRCh38
rs141964431 Chr.19: 45231908-45231908 on Build GRCh38
rs776353432 Chr.19: 45231715-45231715 on Build GRCh38
rs759838271 Chr.19: 45231746-45231746 on Build GRCh38
rs201110701 Chr.19: 45231832-45231832 on Build GRCh38
rs925988156 Chr.19: 45232139-45232139 on Build GRCh38
rs531421726 Chr.19: 45231885-45231885 on Build GRCh38
rs767874981 Chr.19: 45231752-45231752 on Build GRCh38
rs764208057 Chr.19: 45231870-45231870 on Build GRCh38
rs142930668 Chr.19: 45232085-45232085 on Build GRCh38
rs756312859 Chr.19: 45231775-45231775 on Build GRCh38
rs780431532 Chr.19: 45231929-45231929 on Build GRCh38
rs767821387 Chr.19: 45231856-45231856 on Build GRCh38
rs879163734 Chr.19: 45231749-45231749 on Build GRCh38
rs769303645 Chr.19: 45231821-45231821 on Build GRCh38
rs138262010 Chr.19: 45231847-45231847 on Build GRCh38
rs757215478 Chr.19: 45231887-45231887 on Build GRCh38
rs781323519 Chr.19: 45231903-45231903 on Build GRCh38
rs751489490 Chr.19: 45231888-45231888 on Build GRCh38
rs937592097 Chr.19: 45231840-45231840 on Build GRCh38
rs973166803 Chr.19: 45232075-45232075 on Build GRCh38
rs371951355 Chr.19: 45232080-45232080 on Build GRCh38
rs780080755 Chr.19: 45231786-45231786 on Build GRCh38
rs766794060 Chr.19: 45231742-45231742 on Build GRCh38
rs879054127 Chr.19: 45231711-45231711 on Build GRCh38
rs769294719 Chr.19: 45231724-45231724 on Build GRCh38
rs777328011 Chr.19: 45231691-45231691 on Build GRCh38
rs917506538 Chr.19: 45232116-45232116 on Build GRCh38
rs1014772219 Chr.19: 45231764-45231764 on Build GRCh38
rs993077531 Chr.19: 45232088-45232088 on Build GRCh38
rs927458428 Chr.19: 45232079-45232079 on Build GRCh38
rs775800697 Chr.19: 45231858-45231858 on Build GRCh38
rs942176532 Chr.19: 45231758-45231758 on Build GRCh38
rs1030691401 Chr.19: 45231990-45231990 on Build GRCh38
rs900373474 Chr.19: 45231952-45231952 on Build GRCh38
rs369069542 Chr.19: 45231712-45231712 on Build GRCh38
rs202064916 Chr.19: 45231846-45231846 on Build GRCh38
rs370356838 Chr.19: 45231798-45231798 on Build GRCh38
rs748020736 Chr.19: 45231820-45231820 on Build GRCh38
rs1036319032 Chr.19: 45232087-45232087 on Build GRCh38
rs768529039 Chr.19: 45231797-45231797 on Build GRCh38
rs772578859 Chr.19: 45231727-45231727 on Build GRCh38
rs761167246 Chr.19: 45231869-45231869 on Build GRCh38
rs778097506 Chr.19: 45231777-45231777 on Build GRCh38
rs1004970657 Chr.19: 45232025-45232025 on Build GRCh38
rs138339429 Chr.19: 45232045-45232045 on Build GRCh38
rs770491427 Chr.19: 45231841-45231841 on Build GRCh38
rs762567119 Chr.19: 45231733-45231733 on Build GRCh38
rs749686468 Chr.19: 45231932-45231932 on Build GRCh38
rs193056445 Chr.19: 45232119-45232119 on Build GRCh38
rs570910837 Chr.19: 45231950-45231950 on Build GRCh38
rs531905475 Chr.19: 45231717-45231717 on Build GRCh38
rs772987838 Chr.19: 45231825-45231825 on Build GRCh38
rs1014595533 Chr.19: 45232036-45232036 on Build GRCh38
rs752876964 Chr.19: 45231771-45231771 on Build GRCh38
rs761585265 Chr.19: 45231718-45231718 on Build GRCh38
rs754938891 Chr.19: 45231897-45231897 on Build GRCh38
rs906459174 Chr.19: 45231705-45231705 on Build GRCh38
rs1042118242 Chr.19: 45232130-45232130 on Build GRCh38
rs562475810 Chr.19: 45231685-45231685 on Build GRCh38
rs746945112 Chr.19: 45231789-45231789 on Build GRCh38
rs79744739 Chr.19: 45231994-45231994 on Build GRCh38

View Full Product Details