The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003655.2 | NP_003646.2 | 3 - 4 | 345 | 63 |
GenBank mRNA | AF013956.1 | - | 3 - 4 | 348 | 63 |
AY390430.1 | - | 3 - 4 | 182 | 63 | |
BC014967.1 | - | 3 - 4 | 201 | 63 | |
BG029608.1 | - | 3 - 4 | 178 | 63 | |
BQ225831.1 | - | - | 345 | 63 | |
EU439707.1 | - | 3 - 4 | 377 | 63 | |
KU178422.1 | - | 3 - 4 | 182 | 63 | |
KU178423.1 | - | 3 - 4 | 182 | 63 |