Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDC14A
Entrez Gene ID
8556
Gene Name
cell division cycle 14A
Gene Aliases
DFNB105, cdc14, hCDC14
Chromosome Location
Chr.1: 100345015 - 100520281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319210.1 NP_001306139.1 5 - 6 875 83
NM_001319211.1 NP_001306140.1 5 - 6 507 83
NM_001319212.1 NP_001306141.1 5 - 6 875 83
NM_003672.3 NP_003663.2 5 - 6 875 83
NM_033312.2 NP_201569.1 5 - 6 875 83
NM_033313.2 NP_201570.1 5 - 6 875 83
XM_005271294.3 XP_005271351.1 5 - 6 415 83
XM_005271296.3 XP_005271353.1 5 - 6 412 83
XM_011542340.2 XP_011540642.1 5 - 6 414 83
XM_011542341.2 XP_011540643.1 4 - 5 355 83
XM_011542346.2 XP_011540648.1 5 - 6 423 83
XM_017002646.1 XP_016858135.1 5 - 6 415 83
XM_017002647.1 XP_016858136.1 5 - 6 956 83
GenBank mRNA AB209857.1 - 2 - 3 304 83
AF000367.1 - 5 - 6 742 83
AF064102.1 - 5 - 6 850 83
AF064103.1 - 5 - 6 840 83
AF122013.1 - 5 - 6 387 83
AK302310.1 - 5 - 6 496 83
AK310292.1 - 4 - 5 393 83
BC038979.1 - 5 - 6 449 83
BC093916.1 - 5 - 6 482 83
BC093918.1 - 5 - 6 482 83
DQ530256.1 - 5 - 6 393 83

View Full Product Details