The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003764.3 | NP_003755.2 | 1 - 2 | 191 | 126 |
GenBank mRNA | AF044309.1 | - | 1 - 2 | 182 | 126 |
BC033519.1 | - | 1 - 2 | 176 | 126 |