The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160302.1 | NP_001153774.1 | 13 - 14 | 1660 | 111 |
NM_003895.3 | NP_003886.3 | 13 - 14 | 1652 | 111 | |
NM_203446.2 | NP_982271.2 | 13 - 14 | 1652 | 111 | |
XM_017028494.1 | XP_016883983.1 | 13 - 14 | 1642 | 111 | |
XM_017028496.1 | XP_016883985.1 | 13 - 14 | 1642 | 111 | |
XM_017028497.1 | XP_016883986.1 | 13 - 14 | 1740 | 111 | |
XM_017028498.1 | XP_016883987.1 | 13 - 14 | 1642 | 111 | |
XM_017028500.1 | XP_016883989.1 | 13 - 14 | 1642 | 111 | |
XM_017028501.1 | XP_016883990.1 | 13 - 14 | 1642 | 111 | |
XM_017028502.1 | XP_016883991.1 | 13 - 14 | 1642 | 111 | |
XM_017028503.1 | XP_016883992.1 | 13 - 14 | 1642 | 111 | |
XM_017028504.1 | XP_016883993.1 | 13 - 14 | 1642 | 111 | |
GenBank mRNA | AB020717.2 | - | 13 - 14 | 1644 | 111 |
AF009039.1 | - | 13 - 14 | 1578 | 111 | |
AF009040.1 | - | 13 - 14 | 1578 | 111 | |
AK307762.1 | - | 13 - 14 | 1642 | 111 |