Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST3GAL5
Entrez Gene ID
8869
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 5
Gene Aliases
SATI, SIAT9, SIATGM3S, ST3GalV
Chromosome Location
Chr.2: 85839148 - 85889034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042437.1 NP_001035902.1 5 - 6 832 73
NM_003896.3 NP_003887.3 5 - 6 977 73
XM_011533143.2 XP_011531445.1 5 - 6 1105 73
XM_017005199.1 XP_016860688.1 6 - 7 1155 73
XM_017005200.1 XP_016860689.1 6 - 7 2049 73
XM_017005201.1 XP_016860690.1 5 - 6 940 73
XM_017005203.1 XP_016860692.1 7 - 8 1613 73
XM_017005204.1 XP_016860693.1 8 - 9 1694 73
XM_017005205.1 XP_016860694.1 8 - 9 1713 73
XM_017005206.1 XP_016860695.1 7 - 8 1491 73
XM_017005207.1 XP_016860696.1 6 - 7 1091 73
XM_017005208.1 XP_016860697.1 8 - 9 1616 73
XM_017005209.1 XP_016860698.1 7 - 8 1273 73
XM_017005210.1 XP_016860699.1 7 - 8 2620 73
XM_017005211.1 XP_016860700.1 6 - 7 1265 73
XM_017005212.1 XP_016860701.1 6 - 7 2003 73
XM_017005213.1 XP_016860702.1 3 - 4 727 73
GenBank mRNA AB018356.1 - 6 - 7 957 73
AF105026.1 - 5 - 6 911 73
AF119415.1 - 6 - 7 876 73
AF119417.1 - 3 - 4 2083 73
AK001340.1 - 5 - 6 934 73
AK127346.1 - 3 - 4 3197 73
AY152815.2 - 5 - 6 832 73
AY359105.1 - 5 - 6 837 73
BC065936.1 - 6 - 7 893 73
KU178447.1 - 4 - 5 764 73

View Full Product Details