The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078645.1 | NP_001072113.1 | 9 - 10 | 1047 | 96 |
NM_001318517.1 | NP_001305446.1 | 9 - 10 | 1044 | 96 | |
NM_001318518.1 | NP_001305447.1 | 9 - 10 | 1044 | 96 | |
NM_003903.3 | NP_003894.3 | 9 - 10 | 1047 | 96 | |
XM_005266206.1 | XP_005266263.1 | 9 - 10 | 1089 | 96 | |
XM_006719992.1 | XP_006720055.1 | 9 - 10 | 1034 | 96 | |
XM_006719993.1 | XP_006720056.1 | 8 - 9 | 982 | 96 | |
XM_011534851.1 | XP_011533153.1 | 9 - 10 | 1089 | 96 | |
XM_011534853.1 | XP_011533155.1 | 9 - 10 | 1089 | 96 | |
XM_017020830.1 | XP_016876319.1 | 9 - 10 | 1089 | 96 | |
XM_017020831.1 | XP_016876320.1 | 9 - 10 | 1089 | 96 | |
XM_017020832.1 | XP_016876321.1 | 8 - 9 | 982 | 96 | |
XM_017020833.1 | XP_016876322.1 | 7 - 8 | 667 | 96 | |
GenBank mRNA | AB209850.1 | - | 6 - 7 | 4076 | 96 |
AK315302.1 | - | 9 - 10 | 878 | 96 | |
BC010875.2 | - | 9 - 10 | 991 | 96 | |
BC017244.1 | - | 9 - 10 | 957 | 96 | |
U18291.1 | - | 10 - 11 | 870 | 96 |