Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHI3L2
Entrez Gene ID
1117
Gene Name
chitinase 3 like 2
Gene Aliases
CHIL2, YKL-39, YKL39
Chromosome Location
Chr.1: 111227659 - 111243440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004000.2 NP_003991.2 1 - 2 113 58
GenBank mRNA BC011460.1 - 1 - 2 51 58
BG698401.1 - 1 - 2 113 58
BT006767.1 - 1 - 2 42 58
U49835.1 - 1 - 2 62 58
U58514.1 - 1 - 2 78 58

View Full Product Details