Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STOM
Entrez Gene ID
2040
Gene Name
stomatin
Gene Aliases
BND7, EPB7, EPB72
Chromosome Location
Chr.9: 121338988 - 121370304 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004099.5 NP_004090.4 6 - 7 778 85
NR_073037.1 7 - 8 908 85
GenBank mRNA AK302977.1 - 6 - 7 704 85
AK304449.1 - 7 - 8 852 85
AK307999.1 - 6 - 7 721 85
BC010703.1 - 6 - 7 700 85
CR542100.1 - 6 - 7 661 85
M81635.1 - 6 - 7 682 85
X60067.1 - 6 - 7 661 85

View Full Product Details