The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004164.2 | NP_004155.2 | 2 - 3 | 299 | 111 |
GenBank mRNA | AK291978.1 | - | 2 - 3 | 305 | 111 |
AW772327.1 | - | 2 - 3 | 111 | ||
BC069296.1 | - | 2 - 3 | 248 | 111 | |
BC069361.1 | - | 2 - 3 | 248 | 111 | |
BC069396.1 | - | 2 - 3 | 248 | 111 | |
BC069424.1 | - | 2 - 3 | 248 | 111 | |
BC069447.1 | - | 2 - 3 | 248 | 111 | |
BC069469.1 | - | 2 - 3 | 247 | 111 | |
BC069513.1 | - | 2 - 3 | 248 | 111 | |
BC069522.1 | - | 2 - 3 | 248 | 111 | |
BX282005.1 | - | 4 - 5 | 426 | 111 | |
U13831.1 | - | 2 - 3 | 111 |