The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318369.1 | NP_001305298.1 | 9 - 10 | 1761 | 66 |
NM_004211.4 | NP_004202.3 | 10 - 11 | 1900 | 66 | |
XM_017018544.1 | XP_016874033.1 | 6 - 7 | 831 | 66 | |
XM_017018545.1 | XP_016874034.1 | 6 - 7 | 824 | 66 | |
GenBank mRNA | AF085412.1 | - | 10 - 11 | 1908 | 66 |
AF117999.1 | - | 9 - 10 | 1627 | 66 | |
AF142501.1 | - | 9 - 10 | 66 | ||
AF352733.1 | - | 10 - 11 | 1627 | 66 | |
BC096319.3 | - | 10 - 11 | 1706 | 66 | |
BC096320.3 | - | 10 - 11 | 1706 | 66 | |
BC096321.1 | - | 10 - 11 | 1706 | 66 | |
BC096322.3 | - | 9 - 10 | 1567 | 66 |