The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004288.4 | NP_004279.3 | 7 - 8 | 685 | 79 |
XM_017005386.1 | XP_016860875.1 | 7 - 8 | 876 | 79 | |
GenBank mRNA | AF068836.1 | - | 7 - 8 | 646 | 79 |
AK301544.1 | - | 10 - 11 | 778 | 79 | |
AK312753.1 | - | 7 - 8 | 682 | 79 | |
BC036449.2 | - | 7 - 8 | 643 | 79 | |
L06633.1 | - | 7 - 8 | 599 | 79 |