The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004295.3 | NP_004286.2 | 2 - 3 | 301 | 79 |
XM_011525506.2 | XP_011523808.1 | 2 - 3 | 2096 | 79 | |
XM_011525507.2 | XP_011523809.1 | 2 - 3 | 670 | 79 | |
XM_011525508.2 | XP_011523810.1 | 2 - 3 | 387 | 79 | |
GenBank mRNA | AF082185.1 | - | 2 - 3 | 193 | 79 |
AK300804.1 | - | 2 - 3 | 266 | 79 | |
AY937224.1 | - | 2 - 3 | 221 | 79 | |
BC001769.1 | - | 2 - 3 | 285 | 79 | |
CR990949.1 | - | 2 - 3 | 322 | 79 | |
X80200.1 | - | 2 - 3 | 278 | 79 |