Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RGS6
- Entrez Gene ID
-
9628
- Gene Name
- regulator of G-protein signaling 6
- Gene Aliases
-
GAP
- Chromosome Location
-
Chr.14: 71932100 - 72632478 on Build GRCh38
View Full Product Details