The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004308.3 | NP_004299.1 | 3 - 4 | 395 | 57 |
XM_011520095.1 | XP_011518397.1 | 3 - 4 | 832 | 57 | |
XM_017017720.1 | XP_016873209.1 | 2 - 3 | 280 | 57 | |
GenBank mRNA | AK126404.1 | - | 3 - 4 | 832 | 57 |
AK298562.1 | - | 3 - 4 | 342 | 57 | |
AK307296.1 | - | 3 - 4 | 333 | 57 | |
BC018118.1 | - | 3 - 4 | 300 | 57 | |
HY004921.1 | - | 3 - 4 | 396 | 57 | |
U02570.1 | - | 3 - 4 | 302 | 57 | |
Z23024.1 | - | 3 - 4 | 329 | 57 |