The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177924.4 | NP_808592.2 | 1 - 2 | 393 | 107 |
GenBank mRNA | AK098299.1 | - | 1 - 2 | 117 | 107 |
AK222780.1 | - | 1 - 2 | 129 | 107 | |
AK289481.1 | - | 1 - 2 | 294 | 107 | |
BC016481.1 | - | 1 - 2 | 116 | 107 | |
BG714023.1 | - | 1 - 2 | 139 | 107 | |
DA306573.1 | - | 1 - 2 | 393 | 107 | |
U47674.1 | - | 1 - 2 | 97 | 104 | |
U70063.1 | - | 1 - 2 | 98 | 107 |