Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMP1
Entrez Gene ID
1758
Gene Name
dentin matrix acidic phosphoprotein 1
Gene Aliases
ARHP, ARHR, DMP-1
Chromosome Location
Chr.4: 87650302 - 87664361 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004407.3 NP_004398.1 5 - 6 287 88
XM_011531705.2 XP_011530007.1 5 - 6 1162 88
GenBank mRNA BC130581.1 - 4 - 5 209 88
BC132865.1 - 4 - 5 209 88
U89012.1 - 5 - 6 282 88

View Full Product Details