The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287742.1 | NP_001274671.1 | 5 - 6 | 1095 | 90 |
NM_001287743.1 | NP_001274672.1 | 4 - 5 | 897 | 90 | |
NM_001287744.1 | NP_001274673.1 | 3 - 4 | 724 | 90 | |
NM_001287745.1 | NP_001274674.1 | 4 - 5 | 803 | 90 | |
NM_001287747.1 | NP_001274676.1 | 3 - 4 | 760 | 90 | |
NM_001287748.1 | NP_001274677.1 | 3 - 4 | 422 | 90 | |
NM_001287749.1 | NP_001274678.1 | 3 - 4 | 660 | 90 | |
NM_001287750.1 | NP_001274679.1 | 2 - 3 | 662 | 90 | |
NM_001287751.1 | NP_001274680.1 | 2 - 3 | 534 | 90 | |
NM_004462.4 | NP_004453.3 | 3 - 4 | 606 | 90 | |
XM_017013217.1 | XP_016868706.1 | 5 - 6 | 1387 | 90 | |
GenBank mRNA | AK057726.1 | - | 3 - 4 | 724 | 90 |
AK293545.1 | - | 3 - 4 | 660 | 90 | |
AK297868.1 | - | 2 - 3 | 605 | 90 | |
AK300059.1 | - | 2 - 3 | 424 | 90 | |
AK300245.1 | - | 3 - 4 | 383 | 90 | |
AK311246.1 | - | 2 - 3 | 373 | 90 | |
AK311362.1 | - | 3 - 4 | 792 | 90 | |
AK315993.1 | - | 4 - 5 | 673 | 90 | |
AK316033.1 | - | 3 - 4 | 627 | 90 | |
AK316182.1 | - | 2 - 3 | 622 | 90 | |
AK316351.1 | - | 3 - 4 | 387 | 90 | |
AK316531.1 | - | 3 - 4 | 372 | 90 | |
AK316534.1 | - | 2 - 3 | 534 | 90 | |
BC003573.1 | - | 3 - 4 | 453 | 90 | |
BC009251.2 | - | 3 - 4 | 453 | 90 | |
BC029641.1 | - | 3 - 4 | 416 | 90 | |
BC034440.1 | - | 3 - 4 | 416 | 90 | |
BG752446.1 | - | - | 470 | 90 | |
BT006704.1 | - | 3 - 4 | 384 | 90 | |
CR457033.1 | - | 3 - 4 | 384 | 90 | |
DC387479.1 | - | 3 - 4 | 421 | 90 | |
L06070.1 | - | 3 - 4 | 393 | 90 | |
L06105.1 | - | 3 - 4 | 428 | 90 | |
S76822.1 | - | 3 - 4 | 449 | 90 | |
X69141.1 | - | 3 - 4 | 475 | 90 |