Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRG1
Entrez Gene ID
2483
Gene Name
FSHD region gene 1
Gene Aliases
FRG1A, FSG1
Chromosome Location
Chr.4: 189940819 - 189963204 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004477.2 NP_004468.1 5 - 6 630 65
GenBank mRNA AK291890.1 - 5 - 6 604 65
BC053997.1 - 5 - 6 587 65
L76159.1 - 5 - 6 630 65

Gene Symbol
FRG1BP
Entrez Gene ID
284802
Gene Name
FSHD region gene 1 family member B, pseudogene
Gene Aliases
C20orf80, FRG1B, bA348I14.2
Chromosome Location
Chr.20: 30377203 - 30399331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003579.1 4 - 5 535 65
GenBank mRNA BC062743.1 - 5 - 6 754 65
BC095491.1 - 4 - 5 513 65

View Full Product Details