The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001015002.1 | NP_001015002.1 | 6 - 7 | 690 | 88 |
NM_001031803.1 | NP_001026973.1 | 6 - 7 | 690 | 88 | |
NM_004524.2 | NP_004515.2 | 6 - 7 | 690 | 88 | |
XM_011524801.1 | XP_011523103.1 | 6 - 7 | 673 | 88 | |
XM_011524802.1 | XP_011523104.1 | 6 - 7 | 661 | 88 | |
XM_011524807.2 | XP_011523109.1 | 6 - 7 | 778 | 88 | |
XM_017024625.1 | XP_016880114.1 | 6 - 7 | 661 | 88 | |
XM_017024626.1 | XP_016880115.1 | 6 - 7 | 661 | 88 | |
XM_017024627.1 | XP_016880116.1 | 6 - 7 | 661 | 88 | |
XM_017024628.1 | XP_016880117.1 | 6 - 7 | 661 | 88 | |
XM_017024629.1 | XP_016880118.1 | 6 - 7 | 661 | 88 | |
XM_017024630.1 | XP_016880119.1 | 6 - 7 | 661 | 88 | |
XM_017024631.1 | XP_016880120.1 | 6 - 7 | 661 | 88 | |
GenBank mRNA | AK126687.1 | - | 3 - 4 | 713 | 88 |
AK315126.1 | - | 6 - 7 | 640 | 88 | |
BC006503.2 | - | 7 - 8 | 748 | 88 | |
BC010879.1 | - | 6 - 7 | 691 | 88 | |
BC064994.1 | - | 6 - 7 | 663 | 88 | |
X87342.1 | - | 6 - 7 | 666 | 88 |