The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004525.2 | NP_004516.2 | 34 - 35 | 5862 | 72 |
XM_011511183.2 | XP_011509485.1 | 34 - 35 | 5863 | 72 | |
XM_011511184.2 | XP_011509486.1 | 19 - 20 | 3375 | 72 | |
GenBank mRNA | AY265357.1 | - | 34 - 35 | 5716 | 72 |
AY265358.1 | - | 34 - 35 | 5713 | 72 | |
U33837.1 | - | 34 - 35 | 5753 | 72 |