Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A4
Entrez Gene ID
9122
Gene Name
solute carrier family 16 member 4
Gene Aliases
MCT4, MCT5
Chromosome Location
Chr.1: 110362851 - 110391970 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201546.1 NP_001188475.1 7 - 8 1444 87
NM_001201547.1 NP_001188476.1 7 - 8 1455 87
NM_001201549.1 NP_001188478.1 7 - 8 1084 87
NM_001319220.1 NP_001306149.1 6 - 7 951 87
NM_004696.2 NP_004687.1 8 - 9 1588 87
XM_005271317.4 XP_005271374.1 9 - 10 2606 87
XM_006711033.3 XP_006711096.1 6 - 7 2197 87
GenBank mRNA AK129985.1 - 8 - 9 1532 87
AK223307.1 - 8 - 9 1532 87
AK290720.1 - 7 - 8 1388 87
AK295946.1 - 7 - 8 1421 87
AK313735.1 - 8 - 9 1550 87
AL833619.1 - 7 - 8 1385 87
BC021664.1 - 7 - 8 1054 87
BX647948.1 - 6 - 7 917 87
U59185.1 - 8 - 9 1520 87

View Full Product Details