The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004952.4 | NP_004943.1 | 1 - 2 | 202 | 98 |
GenBank mRNA | AK296030.1 | - | 1 - 2 | 186 | 98 |
AK316248.1 | - | 1 - 2 | 165 | 98 | |
BC017722.1 | - | 1 - 2 | 202 | 98 | |
BC110406.1 | - | 1 - 2 | 192 | 98 | |
L37360.1 | - | 1 - 2 | 150 | 98 | |
U14187.1 | - | 1 - 2 | 189 | 98 |