The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267040.1 | NP_001253969.1 | 3 - 4 | 390 | 101 |
NM_004973.3 | NP_004964.2 | 3 - 4 | 890 | 101 | |
XM_011514579.2 | XP_011512881.1 | 5 - 6 | 1034 | 101 | |
XM_011514584.2 | XP_011512886.1 | 1 - 2 | 144 | 101 | |
XM_017010833.1 | XP_016866322.1 | 5 - 6 | 1034 | 101 | |
GenBank mRNA | AK292861.1 | - | 3 - 4 | 736 | 101 |
AK299349.1 | - | 2 - 3 | 304 | 101 | |
AK303610.1 | - | 3 - 4 | 333 | 101 | |
BC046246.1 | - | 3 - 4 | 607 | 101 | |
U57592.1 | - | 3 - 4 | 569 | 101 |