The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272073.1 | NP_001259002.1 | 5 - 6 | 880 | 74 |
NM_005051.2 | NP_005042.1 | 5 - 6 | 913 | 74 | |
NR_073590.1 | 5 - 6 | 864 | 74 | ||
XM_017006965.1 | XP_016862454.1 | 5 - 6 | 593 | 74 | |
GenBank mRNA | AF130067.1 | - | 1 - 2 | 53 | 74 |
AK056815.1 | - | 5 - 6 | 492 | 74 | |
AK222510.1 | - | 5 - 6 | 541 | 74 | |
AK290523.1 | - | 5 - 6 | 541 | 74 | |
AK293260.1 | - | 4 - 5 | 440 | 74 | |
AK297987.1 | - | 4 - 5 | 387 | 74 | |
AK301559.1 | - | 5 - 6 | 509 | 74 | |
AK302867.1 | - | 5 - 6 | 910 | 74 | |
BC000394.2 | - | 5 - 6 | 522 | 74 | |
BC001567.1 | - | 5 - 6 | 520 | 74 | |
BC001772.1 | - | 1 - 2 | 48 | 74 | |
BC016634.1 | - | 4 - 5 | 345 | 74 | |
BC029739.1 | - | 5 - 6 | 523 | 74 | |
X76013.1 | - | 5 - 6 | 522 | 74 |