The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032279.1 | NP_001027450.1 | 4 - 5 | 494 | 74 |
NM_005133.2 | NP_005124.1 | 4 - 5 | 498 | 74 | |
GenBank mRNA | AF121951.1 | - | 4 - 5 | 498 | 74 |
AL713740.1 | - | 3 - 4 | 1193 | 74 | |
BC052622.1 | - | 4 - 5 | 455 | 74 | |
BC093726.1 | - | 4 - 5 | 494 | 74 | |
BC093728.1 | - | 4 - 5 | 493 | 74 | |
Y13835.2 | - | 4 - 5 | 467 | 74 |