Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC12A6
Entrez Gene ID
9990
Gene Name
solute carrier family 12 member 6
Gene Aliases
ACCPN, KCC3, KCC3A, KCC3B
Chromosome Location
Chr.15: 34229996 - 34338064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042494.1 NP_001035959.1 13 - 14 1996 103
NM_001042495.1 NP_001035960.1 13 - 14 1671 103
NM_001042496.1 NP_001035961.1 13 - 14 2062 103
NM_001042497.1 NP_001035962.1 11 - 12 1765 103
NM_005135.2 NP_005126.1 12 - 13 1635 103
NM_133647.1 NP_598408.1 12 - 13 1810 103
XM_006720793.3 XP_006720856.1 11 - 12 2545 103
XM_011522269.2 XP_011520571.1 12 - 13 2692 103
GenBank mRNA AF105366.1 - 12 - 13 1810 103
AF108831.1 - 12 - 13 1544 103
AF116242.1 - 12 - 13 1646 103
AF477977.1 - 13 - 14 2003 103
AF531258.1 - 11 - 12 1765 103
AF531259.1 - 13 - 14 2003 103
AF531260.1 - 13 - 14 1641 103
AK128133.1 - 11 - 12 1318 103
AK292550.1 - 13 - 14 1749 103
AK315283.1 - 12 - 13 1607 103
AL117500.1 - 9 - 10 1238 103
BC070107.1 - 13 - 14 1666 103
BC126241.1 - 12 - 13 1543 103
BC126243.1 - 12 - 13 1543 103
BX648195.1 - 12 - 13 2752 103
DQ138323.1 - 13 - 14 2061 103

View Full Product Details