The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005236.2 | NP_005227.1 | 4 - 5 | 803 | 93 |
XM_011522424.2 | XP_011520726.1 | 5 - 6 | 960 | 93 | |
GenBank mRNA | AK289726.1 | - | 4 - 5 | 802 | 93 |
AK308341.1 | - | 4 - 5 | 769 | 93 | |
BC020741.1 | - | 4 - 5 | 774 | 93 | |
BC142631.1 | - | 4 - 5 | 803 | 93 | |
U64315.1 | - | 4 - 5 | 776 | 93 |