Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYB
Entrez Gene ID
4602
Gene Name
MYB proto-oncogene, transcription factor
Gene Aliases
Cmyb, c-myb, c-myb_CDS, efg
Chromosome Location
Chr.6: 135180981 - 135219173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130172.1 NP_001123644.1 14 - 15 1998 96
NM_001130173.1 NP_001123645.1 15 - 16 2370 96
NM_001161656.1 NP_001155128.1 15 - 16 2361 96
NM_001161657.1 NP_001155129.1 13 - 14 1752 96
NM_001161658.1 NP_001155130.1 15 - 16 2322 96
NM_001161659.1 NP_001155131.1 13 - 14 1896 96
NM_001161660.1 NP_001155132.1 13 - 14 1902 96
NM_005375.3 NP_005366.2 14 - 15 2007 96
NR_134958.1 15 - 16 2129 96
NR_134959.1 15 - 16 2120 96
NR_134960.1 15 - 16 2063 96
NR_134961.1 15 - 16 2190 96
NR_134962.1 14 - 15 1908 96
NR_134963.1 14 - 15 2149 96
NR_134964.1 15 - 16 2054 96
NR_134965.1 16 - 17 2185 96
GenBank mRNA AF104863.1 - 14 - 15 1885 96
AJ606317.1 - 15 - 16 2129 96
AJ606318.1 - 15 - 16 2190 96
AJ606319.1 - 15 - 16 2370 96
AJ606320.1 - 14 - 15 1998 96
AJ606321.1 - 15 - 16 2120 96
AJ606322.1 - 15 - 16 2063 96
AJ606323.1 - 15 - 16 2264 96
AJ616235.1 - 14 - 15 1922 96
AY787443.1 - 13 - 14 1697 96
AY787444.1 - 12 - 13 1515 96
AY787445.1 - 14 - 15 1858 96
AY787446.1 - 14 - 15 1709 96
AY787447.1 - 13 - 14 1703 96
AY787448.1 - 13 - 14 1553 96
AY787449.1 - 14 - 15 1609 96
AY787450.1 - 15 - 16 1864 96
AY787451.1 - 14 - 15 1950 96
AY787452.1 - 14 - 15 2067 96
AY787453.1 - 15 - 16 2064 96
AY787454.1 - 15 - 16 1930 96
AY787455.1 - 16 - 17 2302 96
AY787456.1 - 16 - 17 1986 96
AY787457.1 - 16 - 17 1901 96
AY787458.1 - 15 - 16 1855 96
AY787459.1 - 15 - 16 1921 96
AY787460.1 - 16 - 17 2284 96
AY787461.1 - 15 - 16 2162 96
AY787462.1 - 15 - 16 2311 96
AY787463.1 - 15 - 16 2077 96
AY787464.1 - 15 - 16 1991 96
AY787465.1 - 15 - 16 2405 96
AY787466.1 - 16 - 17 2047 96
AY787467.1 - 15 - 16 2171 96
AY787468.1 - 15 - 16 2123 96
AY787469.1 - 16 - 17 2179 96
AY787470.1 - 14 - 15 1723 96
AY787471.1 - 15 - 16 1779 96
AY787472.1 - 15 - 16 1906 96
AY787473.1 - 15 - 16 2320 96
AY787474.1 - 15 - 16 2086 96
AY787475.1 - 14 - 15 1808 96
BC035996.1 - 8 - 9 1038 96
BC064955.1 - 14 - 15 1959 96
M13665.1 - 12 - 13 1670 96
M15024.1 - 14 - 15 1921 96
X52125.1 - 14 - 15 1996 96
X52126.1 - 14 - 15 1924 96

View Full Product Details