The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005381.2 | NP_005372.2 | 1 - 2 | 158 | 71 |
GenBank mRNA | AK000221.1 | - | 1 - 2 | 116 | 71 |
AK091742.1 | - | 1 - 2 | 148 | 71 | |
AK095897.1 | - | 1 - 2 | 148 | 71 | |
AK122805.1 | - | 1 - 2 | 148 | 71 | |
AK127608.1 | - | - | 165 | 71 | |
BC002343.2 | - | 1 - 2 | 164 | 71 | |
BC006494.2 | - | 1 - 2 | 164 | 71 | |
BC006516.2 | - | 1 - 2 | 164 | 71 | |
BM761058.1 | - | 1 - 2 | 158 | 71 |