The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005468.2 | NP_005459.2 | 4 - 5 | 623 | 76 |
XM_011544706.2 | XP_011543008.1 | 4 - 5 | 943 | 76 | |
XM_011544707.2 | XP_011543009.1 | 5 - 6 | 591 | 76 | |
XM_011544708.2 | XP_011543010.1 | 4 - 5 | 943 | 76 | |
XM_011544709.2 | XP_011543011.1 | 4 - 5 | 963 | 76 | |
XM_011544710.2 | XP_011543012.1 | 4 - 5 | 960 | 76 | |
XM_011544711.2 | XP_011543013.1 | 4 - 5 | 965 | 76 | |
XM_011544712.2 | XP_011543014.1 | 4 - 5 | 966 | 76 | |
GenBank mRNA | AJ012371.1 | - | 4 - 5 | 623 | 76 |