The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005588.2 | NP_005579.2 | 13 - 14 | 2130 | 75 |
XM_011514628.1 | XP_011512930.1 | 12 - 13 | 2187 | 75 | |
GenBank mRNA | AF478685.1 | - | 13 - 14 | 2098 | 75 |
AK225212.1 | - | 13 - 14 | 2120 | 75 | |
AK290282.1 | - | 13 - 14 | 2098 | 75 | |
AK315246.1 | - | 13 - 14 | 2102 | 75 | |
AK315840.1 | - | 13 - 14 | 2100 | 75 | |
BC131714.1 | - | 13 - 14 | 2101 | 75 | |
BC143650.1 | - | 11 - 12 | 2269 | 75 | |
BC143651.1 | - | 12 - 13 | 2180 | 75 | |
M82962.1 | - | 13 - 14 | 2097 | 75 |