The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005655.3 | NP_005646.1 | 1 - 2 | 336 | 71 |
GenBank mRNA | AK312894.1 | - | 1 - 2 | 102 | 71 |
BC095399.1 | - | 1 - 2 | 157 | 71 | |
BI560615.1 | - | 1 - 2 | 194 | 71 | |
DA720479.1 | - | 1 - 2 | 336 | 71 | |
U21847.1 | - | 1 - 2 | 121 | 71 |