Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSCA
Entrez Gene ID
8000
Gene Name
prostate stem cell antigen
Gene Aliases
PRO232
Chromosome Location
Chr.8: 142670308 - 142682727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005672.4 NP_005663.2 1 - 2 107 82
GenBank mRNA AF043498.1 - 1 - 2 68 82
AJ297436.1 - 1 - 2 70 82
AY358912.1 - 1 - 2 40 82
BC023582.2 - 1 - 2 57 82
BC048808.1 - 1 - 2 57 82
BC065183.1 - 1 - 2 57 82
BM768967.1 - 1 - 2 108 82
CB993163.1 - 1 - 2 134 82

View Full Product Details