The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195001.1 | NP_001181930.1 | 1 - 2 | 203 | 53 |
NM_005704.4 | NP_005695.3 | 1 - 2 | 203 | 53 | |
NM_133177.3 | NP_573438.3 | 1 - 2 | 203 | 53 | |
NM_133178.3 | NP_573439.2 | 1 - 2 | 203 | 53 | |
XM_006710269.2 | XP_006710332.1 | 1 - 2 | 178 | 53 | |
XM_016999992.1 | XP_016855481.1 | 1 - 2 | 178 | 53 | |
XM_016999993.1 | XP_016855482.1 | 1 - 2 | 178 | 53 | |
XM_016999994.1 | XP_016855483.1 | 1 - 2 | 178 | 53 | |
GenBank mRNA | BC146655.1 | - | 1 - 2 | 170 | 53 |
U71075.1 | - | 1 - 2 | 80 | 53 | |
U73727.1 | - | 1 - 2 | 194 | 53 | |
X97198.1 | - | 1 - 2 | 206 | 53 |