The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005714.1 | NP_005705.1 | 1 - 2 | 547 | 65 |
NM_033347.1 | NP_203133.1 | 1 - 2 | 547 | 65 | |
NM_033348.1 | NP_203134.1 | 1 - 2 | 547 | 65 | |
NM_033455.1 | NP_258416.1 | 1 - 2 | 547 | 65 | |
GenBank mRNA | AF110522.1 | - | 1 - 2 | 547 | 65 |
AF110523.1 | - | 1 - 2 | 547 | 65 | |
AF110524.1 | - | 1 - 2 | 547 | 65 | |
BC103809.1 | - | 1 - 2 | 360 | 65 | |
BC103810.2 | - | 1 - 2 | 359 | 65 | |
BC103811.2 | - | 1 - 2 | 359 | 65 |