The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005723.3 | NP_005714.2 | 4 - 5 | 891 | 79 |
XM_005262680.1 | XP_005262737.1 | 3 - 4 | 744 | 79 | |
GenBank mRNA | AF065389.1 | - | 4 - 5 | 803 | 79 |
AK126702.1 | - | 1 - 2 | 3530 | 79 | |
AK295385.1 | - | 4 - 5 | 485 | 79 | |
AK315723.1 | - | 4 - 5 | 858 | 79 | |
BC009704.1 | - | 4 - 5 | 789 | 79 | |
CR541809.1 | - | 4 - 5 | 456 | 79 | |
CR541829.1 | - | 4 - 5 | 456 | 79 |