The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251973.1 | NP_001238902.1 | 3 - 4 | 600 | 80 |
NM_001251974.1 | NP_001238903.1 | 3 - 4 | 553 | 80 | |
NM_005822.3 | NP_005813.2 | 2 - 3 | 675 | 80 | |
XM_011514226.1 | XP_011512528.1 | 3 - 4 | 642 | 80 | |
GenBank mRNA | AK090990.1 | - | 3 - 4 | 524 | 80 |
AK312392.1 | - | 2 - 3 | 382 | 80 | |
AY034085.1 | - | 3 - 4 | 507 | 80 | |
AY034086.1 | - | 3 - 4 | 520 | 80 | |
BC038509.1 | - | 2 - 3 | 458 | 80 | |
D83407.1 | - | 2 - 3 | 451 | 80 | |
DA409014.1 | - | 2 - 3 | 308 | 80 |