The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005843.5 | NP_005834.4 | 8 - 9 | 1154 | 94 |
GenBank mRNA | AF042273.1 | - | 8 - 9 | 913 | 94 |
AF042274.1 | - | 8 - 9 | 913 | 94 | |
AK292265.1 | - | 8 - 9 | 849 | 94 | |
AK292847.1 | - | 8 - 9 | 891 | 94 | |
AK310863.1 | - | 8 - 9 | 891 | 94 | |
BC028740.2 | - | 8 - 9 | 1154 | 94 |