Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CWC27
Entrez Gene ID
10283
Gene Name
CWC27 spliceosome associated protein homolog
Gene Aliases
NY-CO-10, SDCCAG-10, SDCCAG10
Chromosome Location
Chr.5: 64768918 - 65018763 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005869.3 NP_005860.2 13 - 14 1484 103
GenBank mRNA AF039692.1 - 13 - 14 1436 103
AF039693.1 - 9 - 10 875 103
AK225105.1 - 13 - 14 1451 103
AK308223.1 - 13 - 14 1454 103
AK312954.1 - 13 - 14 1454 103
AY358569.1 - 13 - 14 1445 103

View Full Product Details