The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005957.4 | NP_005948.3 | 4 - 5 | 820 | 95 |
XM_005263458.3 | XP_005263515.1 | 4 - 5 | 1657 | 95 | |
XM_005263460.4 | XP_005263517.1 | 4 - 5 | 1275 | 95 | |
XM_005263462.4 | XP_005263519.1 | 4 - 5 | 708 | 95 | |
XM_005263463.3 | XP_005263520.1 | 4 - 5 | 837 | 95 | |
XM_011541495.2 | XP_011539797.1 | 4 - 5 | 1654 | 95 | |
XM_011541496.2 | XP_011539798.1 | 4 - 5 | 1657 | 95 | |
XM_017001328.1 | XP_016856817.1 | 4 - 5 | 1657 | 95 | |
GenBank mRNA | AB209113.1 | - | 3 - 4 | 858 | 95 |
AJ237672.1 | - | 3 - 4 | 720 | 95 | |
AK312907.1 | - | 4 - 5 | 681 | 95 | |
BC053509.1 | - | 4 - 5 | 642 | 95 |