Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFS1
Entrez Gene ID
7466
Gene Name
wolframin ER transmembrane glycoprotein
Gene Aliases
CTRCT41, WFRS, WFS, WFSL
Chromosome Location
Chr.4: 6260368 - 6303265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006005.3 NP_005996.2 1 - 2 167 51
GenBank mRNA AF084481.1 - 1 - 2 167 51
AK312897.1 - 1 - 2 96 51
Y18064.1 - 1 - 2 155 51

View Full Product Details