The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079864.2 | NP_001073333.1 | 12 - 13 | 1821 | 89 |
NM_001206901.1 | NP_001193830.1 | 12 - 13 | 1824 | 89 | |
NM_001206902.1 | NP_001193831.1 | 11 - 12 | 1633 | 89 | |
NM_006024.6 | NP_006015.4 | 12 - 13 | 1821 | 89 | |
XM_005249900.2 | XP_005249957.1 | 12 - 13 | 1821 | 89 | |
XM_017012754.1 | XP_016868243.1 | 12 - 13 | 1821 | 89 | |
GenBank mRNA | AF090891.1 | - | 12 - 13 | 1739 | 89 |
AF268075.1 | - | 11 - 12 | 1639 | 89 | |
AK000098.1 | - | 12 - 13 | 1734 | 89 | |
AK296720.1 | - | 11 - 12 | 1582 | 89 | |
AK299729.1 | - | 12 - 13 | 1824 | 89 | |
AK314292.1 | - | 12 - 13 | 1791 | 89 | |
AL136586.1 | - | 12 - 13 | 1727 | 89 | |
BC050358.2 | - | 12 - 13 | 1706 | 89 | |
CR457056.1 | - | 11 - 12 | 1639 | 89 | |
CR533556.1 | - | 11 - 12 | 1639 | 89 | |
U33821.2 | - | 12 - 13 | 1722 | 89 |